Få mere at vide
N-Desmethyl Prochlorperazine-d8 Dimaleate, TRC
Artikelnummer. 30361761
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30361761 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361761 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2920871MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethyl Prochlorperazine-d8 Dimaleate
CAS 1246817-68-4
Molekylær formel C27H22D8ClN3O8S
Mængde 1 mg
Synonym 2-Chloro-10-[3-(1-piperazinyl)propyl]-phenothiazine-d8 Dimaleate
InChI formel InChI=1S/C19H22ClN3S/c20-15-6-7-19-17(14-15)23(16-4-1-2-5-18(16)24-19)11-3-10-22-12-8-21-9-13-22/h1-2,4-7,14,21H,3,8-13H2/i8D2,9D2,12D2,13D2
SMIL ClC1=CC2=C(C=C1)SC3=CC=CC=C3N2CCCN4C([2H])([2H])C([2H])([2H])NC([2H])([2H])C4([2H])[2H]
Molekylvægt (g/mol) 600.11