Få mere at vide
N-Desmethyl Pirenzepine-d8, TRC
Artikelnummer. 30354955
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354955 1 mg
30349472 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354955 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2921321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189860-05-6
Molekylær formel C18H11D8N5O2
Molekylvægt (g/mol) 345.43
Synonym 5,11-Dihydro-11-(1-piperazinyl-d8-acetyl)-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one, LS 822-d8
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(CC(=O)N2c3ccccc3C(=O)Nc4cccnc24)C1([2H])[2H]
Tekniske data
Kemisk navn eller materiale N-Desmethyl Pirenzepine-d8
Mængde 1 mg
InChI formel InChI=1S/C18H19N5O2/c24-16(12-22-10-8-19-9-11-22)23-15-6-2-1-4-13(15)18(25)21-14-5-3-7-20-17(14)23/h1-7,19H,8-12H2,(H,21,25)/i8D2,9D2,10D2,11D2
Formel vægt 345.2041