missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Desmethyl Perazine Dimalonic Acid Salt, TRC
Specifications
Specifications
| Kemisk navn eller materiale | N-Desmethyl Perazine Dimalonic Acid Salt |
| CAS | 60285-35-0 |
| Molekylær formel | C19H23N3S . C6H8O8 |
| Mængde | 25 mg |
| Synonym | 10-[3-(1-Piperazinyl)propyl]-10H-phenothiazine Malonic Acid Salt, 10-(3-Piperazinylpropyl)phenothiazine Malonic Acid Salt, Demethylperazine Malonic Acid Salt, Desmethylperazine Malonic Acid Salt, Norperazine Malonic Acid Salt |
| InChI formel | InChI=1S/C19H23N3S/c1-3-8-18-16(6-1)22(17-7-2-4-9-19(17)23-18)13-5-12-21-14-10-20-11-15-21/h1-4,6-9,20H,5,10-15H2 |
| SMIL | C(CN1CCNCC1)CN2c3ccccc3Sc4ccccc24 |
| Molekylvægt (g/mol) | 325.5 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?