missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl,-N-nitroso-promethazine, TRC

Product Code. 30351552
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30351552 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351552 Supplier Toronto Research Chemicals Supplier No. TRCD39395050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Desmethyl,-N-nitroso-promethazine
CAS 94511-44-1
Molekylær formel C16H17N3OS
Mængde 50 mg
Synonym 10H-Phenothiazine-10-ethanamine, N,α-dimethyl-N-nitroso- (9CI, ACI), N,α-Dimethyl-N-nitroso-10H-phenothiazine-10-ethanamine (ACI), N-Nitroso-N-Desmethyl Promethazine
InChI formel InChI=1S/C16H17N3OS/c1-12(18(2)17-20)11-19-13-7-3-5-9-15(13)21-16-10-6-4-8-14(16)19/h3-10,12H,11H2,1-2H3
SMIL O=NN(C)C(C)CN1C=2C=CC=CC2SC=3C=CC=CC31
IUPAC navn N-(1-(10H-phenothiazin-10-yl)propan-2-yl)-N-methylnitrous amide
Molekylvægt (g/mol) 299.39
Formel vægt 299.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.