Få mere at vide
N-Desmethyl-N-cyclopentyl cis-ent-Tadalafil, TRC
Artikelnummer. 30347223
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30347223 25 mg
30337030 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30347223 Leverandør Toronto Research Chemicals Leverandørnr. TRCB19900525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethyl-N-cyclopentyl cis-ent-Tadalafil
CAS 171488-19-0
Molekylær formel C26 H25 N3 O4
Mængde 25 mg
Synonym N-Desmethyl-N-cyclopentyl cis-ent-Tadalafil
InChI formel InChI=1S/C26H25N3O4/c30-23-13-28(16-5-1-2-6-16)26(31)20-12-18-17-7-3-4-8-19(17)27-24(18)25(29(20)23)15-9-10-21-22(11-15)33-14-32-21/h3-4,7-11,16,20,25,27H,1-2,5-6,12-14H2/t20-,25+/m0/s1
SMIL O=C1CN(C2CCCC2)C(=O)[C@@H]3Cc4c([nH]c5ccccc45)[C@H](N13)c6ccc7OCOc7c6
IUPAC navn (2R,8S)-2-(1,3-benzodioxol-5-yl)-6-cyclopentyl-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
Molekylvægt (g/mol) 443.494
Formel vægt 443.185