Få mere at vide
N-Desmethyl Loperamide-d3, TRC
Artikelnummer. 30358559
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30358559 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358559 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2918421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethyl Loperamide-d3
CAS 1189488-17-2
Molekylær formel C28H28D3ClN2O2
Mængde 1 mg
Synonym N-Desmethyl Loperamide D3
InChI formel InChI=1S/C28H31ClN2O2/c1-30-26(32)28(23-8-4-2-5-9-23,24-10-6-3-7-11-24)18-21-31-19-16-27(33,17-20-31)22-12-14-25(29)15-13-22/h2-15,33H,16-21H2,1H3,(H,30,32)/i1D3
SMIL [2H]C([2H])([2H])NC(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4
Molekylvægt (g/mol) 466.03