Få mere at vide
N-Desmethyl L-Ergothioneine-d6 Methyl Ester, TRC
Artikelnummer. 30356999
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356999 100 mg
30349699 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30356999 Leverandør Toronto Research Chemicals Leverandørnr. TRCD292007100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1795785-97-5
Molekylær formel C9H9D6N3O2S
Molekylvægt (g/mol) 235.34
Synonym (S)-alpha-(Dimethylamino)-2,3-dihydro-2-thioxo-1H-imidazole-4-propanoic-d6 Acid Methyl Ester, N,N-Dimethyl-2-mercapto-L-histidine-d6 Methyl Ester
IUPAC navn methyl (2S)-2-[bis(trideuteriomethyl)amino]-3-(2-sulfanylidene-1,3-dihydroimidazol-4-yl)propanoate
SMIL [2H]C([2H])([2H])N([C@@H](CC1=CNC(=S)N1)C(=O)OC)C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale N-Desmethyl L-Ergothioneine-d6 Methyl Ester
Mængde 100 mg
InChI formel InChI=1S/C9H15N3O2S/c1-12(2)7(8(13)14-3)4-6-5-10-9(15)11-6/h5,7H,4H2,1-3H3,(H2,10,11,15)/t7-/m0/s1/i1D3,2D3
Formel vægt 235.1262