Få mere at vide
N-Desmethyl Droloxifene-d5, TRC
Artikelnummer. 30363308
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30363308 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363308 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2920271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethyl Droloxifene-d5
CAS 1185241-63-7
Molekylær formel C25H22D5NO2
Mængde 1 mg
Synonym 3-[(1E)-1-[4-[2-(Methylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl-d5]-phenol, N-Desmethyldroloxifene-d5, K 106-d5, N-Desmethyl-3-hydroxytamoxifen-d5
InChI formel InChI=1S/C25H27NO2/c1-3-24(19-8-5-4-6-9-19)25(21-10-7-11-22(27)18-21)20-12-14-23(15-13-20)28-17-16-26-2/h4-15,18,26-27H,3,16-17H2,1-2H3/b25-24+/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])\C(=C(\c1ccc(OCCNC)cc1)/c2cccc(O)c2)\c3ccccc3
IUPAC navn 3-[(E)-3,3,4,4,4-pentadeuterio-1-[4-[2-(methylamino)ethoxy]phenyl]-2-phenylbut-1-enyl]phenol
Molekylvægt (g/mol) 378.52
Formel vægt 378.2356