Få mere at vide
N-Desmethyl Diphenhydramine-d3 Hydrochloride, TRC
Artikelnummer. 30350279
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30350279 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350279 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2917822.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethyl Diphenhydramine-d3 Hydrochloride
CAS 1794759-12-8
Molekylær formel C16H17D3ClNO
Mængde 2.5 mg
Synonym 2-(Diphenylmethoxy)-N-(methyl-d3)ethanamine Hydrochloride, 2-(Diphenylmethoxy)-N-(methyl-d3)ethylamine Hydrochloride
InChI formel InChI=1S/C16H19NO.ClH/c1-17-12-13-18-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15;/h2-11,16-17H,12-13H2,1H3;1H/i1D3;
SMIL [2H]C([2H])([2H])NCCOC(C1=CC=CC=C1)C2=CC=CC=C2.Cl
Molekylvægt (g/mol) 280.81