Få mere at vide
N-Desmethyl-4’-hydroxy Tamoxifen, TRC
Artikelnummer. 30365722
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365722 25 mg
30353671 5 mg
30359139 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365722 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29204125MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 170171-12-7
Molekylær formel C25H27NO2
Molekylvægt (g/mol) 373.49
SMIL CC\C(=C(/c1ccccc1)\c2ccc(OCCNC)cc2)\c3ccc(O)cc3
Tekniske data
Kemisk navn eller materiale N-Desmethyl-4'-hydroxy Tamoxifen
Mængde 25 mg
InChI formel InChI=1S/C25H27NO2/c1-3-24(19-9-13-22(27)14-10-19)25(20-7-5-4-6-8-20)21-11-15-23(16-12-21)28-18-17-26-2/h4-16,26-27H,3,17-18H2,1-2H3/b25-24-
Formel vægt 373.2042