Få mere at vide
N-Desmethyl-4'-hydroxy Tamoxifen-d3 (E/Z Mixture), TRC
Artikelnummer. 30366565
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30366565 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366565 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29186710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethyl-4'-hydroxy Tamoxifen-d3 (E/Z Mixture)
CAS 1795139-22-8
Molekylær formel C25H24D3NO2
Mængde 10 mg
Synonym 4-[(1E/Z)-1-[[4-[2-(Methylamino-d3)ethoxy]phenyl]phenylmethylene]propyl]phenol
InChI formel InChI=1S/C25H27NO2/c1-3-24(19-9-13-22(27)14-10-19)25(20-7-5-4-6-8-20)21-11-15-23(16-12-21)28-18-17-26-2/h4-16,26-27H,3,17-18H2,1-2H3/i2D3
SMIL OC(C=C1)=CC=C1/C(CC)=C(C2=CC=C(OCCNC([2H])([2H])[2H])C=C2)/C3=CC=CC=C3
Molekylvægt (g/mol) 376.51