Få mere at vide
N-Desmethyl-4-hydroxy Tamoxifen (approx. 1:1 E/Z Mixture), TRC
Artikelnummer. 30136544
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136544 25 mg
30004316 5 mg
30006013 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136544 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29204325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 110025-28-0
Molekylær formel C25H27NO2
Molekylvægt (g/mol) 373.49
Synonym 4-[1-[4-[2-(Methylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]phenol, (E/Z)-Endoxifen, 4OHNDtam;
SMIL OC1=CC=C(/C(C2=CC=C(OCCNC)C=C2)=C(C3=CC=CC=C3)\CC)C=C1
Tekniske data
Kemisk navn eller materiale N-Desmethyl-4-hydroxy Tamoxifen (E/Z Mixture)
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C25H27NO2/c1-3-24(19-7-5-4-6-8-19)25(20-9-13-22(27)14-10-20)21-11-15-23(16-12-21)28-18-17-26-2/h4-16,26-27H,3,17-18H2,1-2H3/b25-24-
Procent renhed >95%