Få mere at vide
N-Desisopropyl Metoprolol-d5, TRC
Artikelnummer. 30366401
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30366401 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366401 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29024250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desisopropyl Metoprolol-d5
CAS 1794979-77-3
Molekylær formel C12H14D5NO3
Mængde 50 mg
Synonym 1-Amino-3-[4-(2-methoxyethyl)phenoxy]-2-propanol-d5, H 98/52-d5
InChI formel InChI=1S/C12H19NO3/c1-15-7-6-10-2-4-12(5-3-10)16-9-11(14)8-13/h2-5,11,14H,6-9,13H2,1H3/i8D2,9D2,11D
SMIL NC([2H])([2H])C(O)([2H])C([2H])([2H])OC1=CC=C(C=C1)CCOC
IUPAC navn 1-amino-3-(4-(2-methoxyethyl)phenoxy)propan-1,1,2,3,3-d5-2-ol
Molekylvægt (g/mol) 230.31
Formel vægt 230.17