Få mere at vide
N-Desethylvardenafil, Mikromol™
Artikelnummer. 30049450
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049450 25 mg
17999214 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049450 Leverandør Mikromol Leverandørnr. MM1569.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Desethylvardenafil
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desethylvardenafil
CAS 448184-46-1
Analyt- eller komponentnavne N-Desethylvardenafil
Forsendelsestilstand Room Temperature
Molekylær formel C21 H28 N6 O4 S
Certificeringer/compliance ISO 9001
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-[2-ethoxy-5-(1-piperazinylsulfonyl)phenyl]-5-methyl-7-propyl-, Piperazine, 1-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), N-Desethylvardenafil, Vardenafil Related Compound E
InChI formel InChI=1S/C21H28N6O4S/c1-4-6-18-23-14(3)19-21(28)24-20(25-27(18)19)16-13-15(7-8-17(16)31-5-2)32(29,30)26-11-9-22-10-12-26/h7-8,13,22H,4-6,9-12H2,1-3H3,(H,24,25,28)
SMIL CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CCNCC4
IUPAC navn 2-(2-ethoxy-5-piperazin-1-ylsulfonylphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
Molekylvægt (g/mol) 460.55
Mængde 25 mg
Formel vægt 460.1893 g/mol