Få mere at vide
N-Desethylvardenafil, LoGiCal
Artikelnummer. 30461030
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461030 10 mg
1 options
Artikelnummer. 30461030 Leverandør LoGiCal Leverandørnr. LGCFOR1569.02
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desethylvardenafil
CAS 448184-46-1
Analyt- eller komponentnavne N-Desethylvardenafil
Forsendelsestilstand Room Temperature
Molekylær formel C21 H28 N6 O4 S
Certificeringer/compliance ISO 9001
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-[2-ethoxy-5-(1-piperazinylsulfonyl)phenyl]-5-methyl-7-propyl-, Piperazine, 1-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), N-Desethylvardenafil, Vardenafil Related Compound E
InChI formel InChI=1S/C21H28N6O4S/c1-4-6-18-23-14(3)19-21(28)24-20(25-27(18)19)16-13-15(7-8-17(16)31-5-2)32(29,30)26-11-9-22-10-12-26/h7-8,13,22H,4-6,9-12H2,1-3H3,(H,24,25,28)
Løsningstype Neat
SMIL CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CCNCC4
IUPAC navn 2-(2-ethoxy-5-piperazin-1-ylsulfonylphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
Industry Type Clinical Testing
Molekylvægt (g/mol) 460.55
Mængde 10 mg
Formel vægt 460.1893 g/mol