Få mere at vide
N-Desethyl Vardenafil-d8, TRC
Artikelnummer. 30349025
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30349025 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349025 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28995210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desethyl Vardenafil-d8
CAS 1184985-26-9
Molekylær formel C21H20D8N6O4S
Mængde 10 mg
Synonym 1-[[3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]-piperazine-d8
InChI formel InChI=1S/C21H28N6O4S/c1-4-6-18-23-14(3)19-21(28)24-20(25-27(18)19)16-13-15(7-8-17(16)31-5-2)32(29,30)26-11-9-22-10-12-26/h7-8,13,22H,4-6,9-12H2,1-3H3,(H,24,25,28)/i9D2,10D2,11D2,12D2
SMIL CCOC(C(C1=NN2C(C(N1)=O)=C(C)N=C2CCC)=C3)=CC=C3S(N4C([2H])([2H])C([2H])([2H])NC([2H])([2H])C4([2H])[2H])(=O)=O
Molekylvægt (g/mol) 468.6