Få mere at vide
N-Desethyl Sunitinib-d5, TRC
Artikelnummer. 30355880
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30355880 10 mg
30357293 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30355880 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28965210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217247-62-5
Molekylær formel C20H18D5FN4O2
Molekylvægt (g/mol) 375.45
Synonym N-[2-(Ethylamino-d5)ethyl]-5-[(Z)-(5-fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxamide
SMIL O=C(/C1=C\C2=C(C)C(C(NCCNC([2H])([2H])C([2H])([2H])[2H])=O)=C(C)N2)NC3=C1C=C(F)C=C3
Tekniske data
Kemisk navn eller materiale N-Desethyl Sunitinib-d5
Mængde 10 mg
InChI formel InChI=1S/C20H23FN4O2/c1-4-22-7-8-23-20(27)18-11(2)17(24-12(18)3)10-15-14-9-13(21)5-6-16(14)25-19(15)26/h5-6,9-10,22,24H,4,7-8H2,1-3H3,(H,23,27)(H,25,26)/b15-10-/i1D3,4D2