Få mere at vide
N-Desethyl Milnacipran-d5, TRC
Artikelnummer. 30364384
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30364384 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30364384 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2894021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desethyl Milnacipran-d5
CAS 1217609-30-7
Molekylær formel C13 H13 D5 N2 O
Mængde 1 mg
Synonym N-Desethyl Milnacipran D5
InChI formel InChI=1S/C13H18N2O/c1-2-15-12(16)13(8-11(13)9-14)10-6-4-3-5-7-10/h3-7,11H,2,8-9,14H2,1H3,(H,15,16)/t11-,13+/m1/s1/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])N=C(O)[C@@]1(C[C@@H]1CN)c2ccccc2
Molekylvægt (g/mol) 223.33