Få mere at vide
N-Desethyl Amodiaquine-d5 Dihydrochloride, TRC
Artikelnummer. 30355678
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30355678 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30355678 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28882810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desethylamodiaquine-D5 diHCl
CAS 1216894-33-5
Renhedsgrad noter HPLC
Molekylær formel C18 2H5 H13 Cl N3 O . 2 Cl H
Mængde 10 mg
Synonym Desethylamodiaquine-D5 Dihydrochloride
InChI formel InChI=1S/C18H18ClN3O.2ClH/c1-2-20-11-12-9-14(4-6-18(12)23)22-16-7-8-21-17-10-13(19)3-5-15(16)17;;/h3-10,20,23H,2,11H2,1H3,(H,21,22);2*1H/i1D3,2D2;;
SMIL Cl.Cl.[2H]C([2H])([2H])C([2H])([2H])NCc1cc(Nc2ccnc3cc(Cl)ccc23)ccc1O
IUPAC navn 4-[(7-chloroquinolin-4-yl)amino]-2-[(1,1,2,2,2-pentadeuterioethylamino)methyl]phenol;dihydrochloride
Molekylvægt (g/mol) 405.76
Formel vægt 404.0986
Procent renhed >95