Få mere at vide
N-Descarboxymethyl-N-carboxyethyl Lacosamide (Impurity), TRC
Artikelnummer. 30357449
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30357449 50 mg
30363456 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30357449 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28939050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1318777-56-8
Molekylær formel C14 H20 N2 O3
Molekylvægt (g/mol) 264.32
Synonym N-Descarboxymethyl-N-carboxyethyl Lacosamide (Impurity), Propanamide, 3-methoxy-2-[(1-oxopropyl)amino]-N-(phenylmethyl)-, (2R)- (ACI), (2R)-3-Methoxy-2-[(1-oxopropyl)amino]-N-(phenylmethyl)propanamide (ACI), (R)-N-Benzyl-3-methoxy-2-propionamidopropanamide, (R)-N-benzyl-2-((1-hydroxypropylidene)amino)-3-methoxypropanimidic acid, N-Descarboxymethyl-N-carboxyethyl Lacosamide (Impurity)
IUPAC navn (2R)-N-benzyl-3-methoxy-2-(propanoylamino)propanamide
SMIL CCC(=O)N[C@H](COC)C(=O)NCc1ccccc1
Tekniske data
Kemisk navn eller materiale (2R)-3-Methoxy-2-[(1-oxopropyl)amino]-N-(phenylmethyl)propanamide
Mængde 50 mg
InChI formel InChI=1S/C14H20N2O3/c1-3-13(17)16-12(10-19-2)14(18)15-9-11-7-5-4-6-8-11/h4-8,12H,3,9-10H2,1-2H3,(H,15,18)(H,16,17)/t12-/m1/s1
Formel vægt 264.147