Få mere at vide
N-DesBoc Docetaxel, TRC
Artikelnummer. 30363743
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363743 50 mg
30362347 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363743 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28923550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 133524-69-3
Molekylær formel C38H45NO12
Molekylvægt (g/mol) 707.77
Synonym Benzenepropanoic acid, β-amino-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,6,11-trihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-amino-α-hydroxy-, 12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,6,11-trihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv., 10-Deacetyl-3'-debenzoylpaclitaxel, 10-Deacetylbaccatin III 13-[β(S)-amino-α(R)-hydroxyphenylpropionate], 10-Desacetyl-N-debenzoylpaclitaxel, N-De(tert-butoxycarbonyl)-10-deacetyldocetaxel, N-Debenzoyl-10-deacetylpaclitaxel
SMIL CC(=O)O[C@@]12CO[C@@H]1C[C@H](O)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](N)c6ccccc6)C(=C([C@@H](O)C3=O)C5(C)C)C
Tekniske data
Kemisk navn eller materiale N-DesBoc Docetaxel
Mængde 50 mg
InChI formel InChI=1S/C38H45NO12/c1-19-23(49-34(46)29(43)27(39)21-12-8-6-9-13-21)17-38(47)32(50-33(45)22-14-10-7-11-15-22)30-36(5,31(44)28(42)26(19)35(38,3)4)24(41)16-25-37(30,18-48-25)51-20(2)40/h6-15,23-25,27-30,32,41-43,47H,16-18,39H2,1-5H3/t23-,24-,25+,27-,28+,29+,30-,32-,36+,37-,38+/m0/s1
Formel vægt 707.2942