Få mere at vide
N-Desacetyl 5-Azido Oseltamivir, TRC
Artikelnummer. 30358544
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358544 50 mg
30349631 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30358544 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28841550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 204255-04-9
Molekylær formel C14 H24 N4 O3
Molekylvægt (g/mol) 296.37
Synonym 1-Cyclohexene-1-carboxylic acid, 4-amino-5-azido-3-(1-ethylpropoxy)-, ethyl ester, (3R,4R,5S)-, ethyl (3R,4R,5S)-4-amino-5-azido-3-(1-ethylpropoxy)cyclohexene-1-carboxylate
IUPAC navn ethyl (3R,4R,5S)-4-amino-5-azido-3-pentan-3-yloxycyclohexene-1-carboxylate
SMIL CCOC(=O)C1=C[C@@H](OC(CC)CC)[C@H](N)[C@H](C1)N=[N+]=[N-]
Tekniske data
Kemisk navn eller materiale N-Desacetyl 5-Azido Oseltamivir
Mængde 50 mg
InChI formel InChI=1S/C14H24N4O3/c1-4-10(5-2)21-12-8-9(14(19)20-6-3)7-11(13(12)15)17-18-16/h8,10-13H,4-7,15H2,1-3H3/t11-,12+,13+/m0/s1
Formel vægt 296.1848