Få mere at vide
N-des-succinyl-Sacubitril hydrochloride, TRC
Artikelnummer. 30136938
Skift visning
Mængde:
50 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136938 1 g
30135456 5 g
30007058 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136938 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6015401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 149690-12-0
Molekylær formel C20 H25 N O2 . H Cl
Synonym (2R,4S)-4-Amino-5-(biphenyl-4-yl)-2-methylpentanoic Acid Ethyl Ester Hydrochloride∣(αR,γS)-γ-Amino-α-methyl-[1,1'-biphenyl]-4-pentanoic Acid Ethyl Ester Hydrochloride∣[S-(R*,S*)]-γ-Amino-α-methyl-[1,1'-biphenyl]-4-pentanoic Acid Ethyl Ester Hydrochloride∣Ethyl (2R,4S)-5-([1,1'-Biphenyl]-4-yl)-4-amino-2-methylpentanoate Hydrochloride
Tekniske data
Kemisk navn eller materiale (2R,4S)-4-Amino-5-(biphenyl-4-yl)-2-methylpentanoic Acid Ethyl Ester Hydrochloride
Mængde 1 g
Forsendelsestilstand Room Temperature
Formel vægt 347.17
MDL nummer MFCD27955986
Molekylvægt (g/mol) 347.88
InChI nøgle MYJTZLYRAWUSLB-WSCVZUBPSA-N
SMIL Cl.CCOC(=O)[C@H](C)C[C@H](N)CC1=CC=C(C=C1)C1=CC=CC=C1
IUPAC navn ethyl (2R,4S)-4-amino-5-{[1,1'-biphenyl]-4-yl}-2-methylpentanoate hydrochloride