Få mere at vide
N-Des[2-(1-methyl-2-pyrrolidinyl)ethyl] Udenafil, TRC
Artikelnummer. 30353989
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353989 25 mg
30350051 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30353989 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29265525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 319491-68-4
Molekylær formel C18 H23 N5 O4 S
Molekylvægt (g/mol) 405.4713
Synonym Benzenesulfonamide, 3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxy- (9CI), 3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxybenzenesulfonamide, DA 8164, Udenafil N-desalkyl
IUPAC navn 3-(1-methyl-7-oxo-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxybenzenesulfonamide
SMIL CCCOc1ccc(cc1C2=NC(=O)c3c(N2)c(CCC)nn3C)S(=O)(=O)N
Tekniske data
Kemisk navn eller materiale N-Desalkyludenafil
Mængde 25 mg
InChI formel InChI=1S/C18H23N5O4S/c1-4-6-13-15-16(23(3)22-13)18(24)21-17(20-15)12-10-11(28(19,25)26)7-8-14(12)27-9-5-2/h7-8,10H,4-6,9H2,1-3H3,(H2,19,25,26)(H,20,21,24)
Formel vægt 405.1471