Få mere at vide
N-Depropyl Propafenone-d5 Oxalate Salt, TRC
Artikelnummer. 30364817
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30364817 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30364817 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2886622.5MG
Denne vare kan ikke returneres. Se returpolitik
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Depropyl Propafenone-d5 Oxalate Salt
CAS 1215598-59-6
Molekylær formel C20H18D5NO7
Mængde 2.5 mg
Synonym N-Desalkylpropafenone-d5 Oxalate Salt, N-Despropylpropafenone-d5 Oxalate Salt
InChI formel InChI=1S/C18H21NO3.C2H2O4/c19-12-15(20)13-22-18-9-5-4-8-16(18)17(21)11-10-14-6-2-1-3-7-14;3-1(4)2(5)6/h1-9,15,20H,10-13,19H2;(H,3,4)(H,5,6)/i12D2,13D2,15D;
SMIL [2H]C([2H])(N)C([2H])(O)C([2H])([2H])Oc1ccccc1C(=O)CCc2ccccc2.OC(=O)C(=O)O
IUPAC navn 1-[2-(3-amino-1,1,2,3,3-pentadeuterio-2-hydroxypropoxy)phenyl]-3-phenylpropan-1-one;oxalic acid
Molekylvægt (g/mol) 394.43
Formel vægt 394.1788