Få mere at vide
N-(Deoxyguanosin-8-yl)-2-amino-1-(methyl-d3)-6-phenylimidazo[4,5-Beta]pyridine, TRC
Artikelnummer. 30358196
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30358196 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358196 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2396320.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(Deoxyguanosin-8-yl)-2-amino-1-(methyl-d3)-6-phenylimidazo[4,5-beta]pyridine
CAS 303173-39-9
Molekylær formel C23 2H3 H20 N9 O4
Mængde 0.25 mg
Synonym 2'-Deoxy-8-[[1-(methyl-d3)-6-phenyl-1H-imidazo[4,5-b]pyridin-2-yl]amino]guanosine, dG-C8-PhIP-d3
InChI formel InChI=1S/C23H23N9O4/c1-31-13-7-12(11-5-3-2-4-6-11)9-25-18(13)27-22(31)30-23-26-17-19(28-21(24)29-20(17)35)32(23)16-8-14(34)15(10-33)36-16/h2-7,9,14-16,33-34H,8,10H2,1H3,(H3,24,28,29,35)(H,25,26,27,30)/t14-,15+,16+/m0/s1/i1D3
SMIL [2H]C([2H])([2H])n1c(Nc2nc3C(=O)NC(=Nc3n2[C@H]4C[C@H](O)[C@@H](CO)O4)N)nc5ncc(cc15)c6ccccc6
IUPAC navn 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-8-[[6-phenyl-1-(trideuteriomethyl)imidazo[4,5-b]pyridin-2-yl]amino]-1H-purin-6-one
Molekylvægt (g/mol) 492.51
Formel vægt 492.2061