Få mere at vide
N-Demethylterbinafine Hydrochloride, Mikromol™
Beskrivelse
N-Demethylterbinafine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Demethylterbinafine Hydrochloride |
| CAS | 152830-98-3 |
| Analyt- eller komponentnavne | N-Demethylterbinafine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H23 N . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | N-Demethylterbinafine Hydrochloride |
| InChI formel | InChI=1S/C20H23N.ClH/c1-20(2,3)14-7-4-8-15-21-16-18-12-9-11-17-10-5-6-13-19(17)18;/h4-6,8-13,21H,15-16H2,1-3H3;1H/b8-4+; |
| SMIL | Cl.CC(C)(C)C#C\C=C\CNCc1cccc2ccccc12 |
| IUPAC navn | (E)-6,6-dimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride |
| Vis mere |