missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Demethyl Trimebutine Hydrochloride, TRC

Product Code. 30138521
Change view
Click to view available options
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30138521 25 mg
30008826 5 mg
30011689 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138521 Supplier Toronto Research Chemicals Supplier No. TRCD23134025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 294882-33-0
Molekylær formel C21 H27 N O5 . Cl H
Molekylvægt (g/mol) 409.9
Synonym Benzoic acid, 3,4,5-trimethoxy-, 2-(methylamino)-2-phenylbutyl ester, hydrochloride (1:1), Benzoic acid, 3,4,5-trimethoxy-, 2-(methylamino)-2-phenylbutyl ester, hydrochloride (9CI), (2RS)-2-(Methylamino)-2-phenylbutyl 3,4,5-Trimethoxy-benzoate Hydrochloride, Trimebutine Imp. E (EP) as Hydrochloride, N-Desmethyltrimebutine Hydrochloride
IUPAC navn [2-(methylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate;hydrochloride
SMIL Cl.CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(NC)c2ccccc2

Specifications

Kemisk navn eller materiale N-Demethyl Trimebutine hydrochloride
Mængde 25 mg
InChI formel InChI=1S/C21H27NO5.ClH/c1-6-21(22-2,16-10-8-7-9-11-16)14-27-20(23)15-12-17(24-3)19(26-5)18(13-15)25-4;/h7-13,22H,6,14H2,1-5H3;1H
Formel vægt 409.1656
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.