Få mere at vide
N-Demethyl Olanzapine-d8, TRC
Artikelnummer. 30367145
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30367145 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367145 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2309721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethylolanzapine-d8
CAS 786686-82-6
Molekylær formel C16 2H8 H10 N4 S
Mængde 1 mg
Synonym N-Desmethylolanzapine-D8, 10H-Thieno[2,3-b][1,5]benzodiazepine, 2-methyl-4-(1-piperazinyl-2,2,3,3,5,5,6,6-d8)- (9CI)
InChI formel InChI=1S/C16H18N4S/c1-11-10-12-15(20-8-6-17-7-9-20)18-13-4-2-3-5-14(13)19-16(12)21-11/h2-5,10,17,19H,6-9H2,1H3/i6D2,7D2,8D2,9D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(C2=Nc3ccccc3Nc4sc(C)cc24)C1([2H])[2H]
IUPAC navn 2-methyl-4-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-10H-thieno[2,3-b][1,5]benzodiazepine
Molekylvægt (g/mol) 306.46
Formel vægt 306.1754