Få mere at vide
N-Demethyl Eszopiclone Hydrochloride, TRC
Artikelnummer. 30135209
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135209 50 mg
30008574 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135209 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23093550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 300701-71-7
Molekylær formel C16H16Cl2N6O3
Molekylvægt (g/mol) 411.24
Synonym 1-Piperazinecarboxylic Acid (5S)-6-(5-Chloro-2-pyridinyl)-6,7-dihydro-7-oxo-
SMIL Cl.Clc1ccc(nc1)N2[C@@H](OC(=O)N3CCNCC3)c4nccnc4C2=O
Tekniske data
Kemisk navn eller materiale N-Demethyl Eszopiclone Hydrochloride
Mængde 50 mg
InChI formel InChI=1S/C16H15ClN6O3.ClH/c17-10-1-2-11(21-9-10)23-14(24)12-13(20-4-3-19-12)15(23)26-16(25)22-7-5-18-6-8-22;/h1-4,9,15,18H,5-8H2;1H/t15-;/m0./s1