Få mere at vide
N-Demethyl-3,4-dihydroxytamoxifen Hydrochloride(Z,E mixture), TRC
Artikelnummer. 30356755
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356755 50 mg
30354422 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30356755 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23103550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Demethyl-3,4-dihydroxytamoxifen Hydrochloride (E,Z mixture)
CAS 2125720-11-6
Molekylær formel C25H28ClNO3
Mængde 50 mg
Synonym 4-[(Z,E)-1-[4-[2-(Methylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]-1,2-benzenediol Hydrochloride
InChI formel InChI=1S/C25H27NO3.ClH/c1-3-22(18-7-5-4-6-8-18)25(20-11-14-23(27)24(28)17-20)19-9-12-21(13-10-19)29-16-15-26-2;/h4-14,17,26-28H,3,15-16H2,1-2H3;1H/b25-22+;
SMIL OC(C(O)=C1)=CC=C1/C(C2=CC=C(OCCNC)C=C2)=C(CC)/C3=CC=CC=C3.Cl
IUPAC navn 4-(1-(4-(2-(methylamino)ethoxy)phenyl)-2-phenylbut-1-en-1-yl)benzene-1,2-diol hydrochloride
Molekylvægt (g/mol) 425.95
Formel vægt 425.18