Learn More
n-Decyltrimethoxysilane, 97% Product Code.: 11333866
n-Decyltrimethoxysilane, 97%
Product Code. p-7046961
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11333866
Brand: Thermo Scientific Alfa Aesar L16966.14
Request Bulk or Custom Format
CAS: 5575-48-4 | C13H30O3Si | 262.47 g/mol
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5575-48-4
Molecular Formula C13H30O3Si
Molecular Weight (g/mol) 262.47
MDL Number MFCD00069135
InChI Key KQAHMVLQCSALSX-UHFFFAOYSA-N
Synonym n-decyltrimethoxysilane, decyltrimethoxysilan, trimethoxydecylsilane, decyl trimethoxy silane, silane,decyltrimethoxy, decyl-trimethoxysilane
PubChem CID 2758023
IUPAC Name decyl(trimethoxy)silane
SMILES CCCCCCCCCC[Si](OC)(OC)OC
Specifications
Boiling Point 102°C to 106°C (3.5mmHg)
Melting Point -37°C
Quantity 25 g
Beilstein 8625082
Flash Point 117°C (242°F)
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 262.47
Percent Purity 98%
Refractive Index 1.422
Sensitivity Moisture sensitive
Density 0.9
Chemical Name or Material n-Decyltrimethoxysilane
Show More Show Less
RUO – Research Use Only
Product Content Correction
Product Title
n-Decyltrimethoxysilane, 97% >