Få mere at vide
n-Decyl-1,1,2,2-d4 Alcohol, TRC
Artikelnummer. 30356660
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30356660 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30356660 Leverandør Toronto Research Chemicals Leverandørnr. TRCD21254350MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Decanol-1,1,2,2-D4
CAS 57367-97-2
Molekylær formel C10H18D4O
Mængde 50 mg
Synonym 1-Decan-1,1,2,2-d4-ol (9CI), 1,1,2,2-Tetradeuteriodecan-1-ol, 1-Decanol-1,1,2,2-d4, 1-Decanol-1,1,2,2-D4, n-Decyl-1,1,2,2-d4 alcohol
InChI formel InChI=1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3/i9D2,10D2
SMIL CCCCCCCCC([2H])([2H])C([2H])([2H])O
IUPAC navn decan-1,1,2,2-d4-1-ol
Molekylvægt (g/mol) 162.31
Formel vægt 162.19