Få mere at vide
N-Dechloroethyl Cyclophosphamide-d4, TRC
Artikelnummer. 30352778
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30352778 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352778 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22640210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Dechloroethyl Cyclophosphamide-d4
CAS 1346600-68-7
Molekylær formel C5H8D4ClN2O2P
Mængde 10 mg
Synonym N-(2-Chloroethyl-d4)tetrahydro-2H-1,3,2-oxazaphosphorin-2-amine 2-Oxide, 2-(2-Chloroethylamino-d4)tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, 3-Dechloroethylifosfamide-d4, Asta 4968-d4, Dechloroethylcyclophosphamide-d4, Monochloroethylcyclophosphamide-d4
InChI formel InChI=1S/C5H12ClN2O2P/c6-2-4-8-11(9)7-3-1-5-10-11/h1-5H2,(H2,7,8,9)/i2D2,4D2
SMIL [2H]C([2H])(Cl)C([2H])([2H])NP1(=O)NCCCO1
IUPAC navn N-(2-chloro-1,1,2,2-tetradeuterioethyl)-2-oxo-1,3,2λ5-oxazaphosphinan-2-amine
Molekylvægt (g/mol) 202.61
Formel vægt 202.0576