Få mere at vide
N-Dansyl-D-phenylalanine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Dansyl-D-phenylalanine |
| CAS | 56176-31-9 |
| Molekylær formel | C21H22N2O4S |
| Synonym | N-[[5-(Dimethylamino)-1-naphthalenyl]sulfonyl]-D-phenylalanine, D-Dansylphenylalanine, Dansyl-(R)-phenylalanine, Dansyl-D-Phe, Dansyl-D-phenylalanine, N-Dansyl-D-phenylalanine; |
| InChI formel | InChI=1S/C21H22N2O4S/c1-23(2)19-12-6-11-17-16(19)10-7-13-20(17)28(26,27)22-18(21(24)25)14-15-8-4-3-5-9-15/h3-13,18,22H,14H2,1-2H3,(H,24,25)/t18-/m1/s1 |
| SMIL | CN(C)c1cccc2c(cccc12)S(=O)(=O)N[C@H](Cc3ccccc3)C(=O)O |
| IUPAC navn | (2R)-2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]-3-phenylpropanoic acid |
| Molekylvægt (g/mol) | 398.48 |
| Mængde | 250 mg |
| Forsendelsestilstand | Room Temperature |