missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Cyclohexyl-4-pentenamide, TRC

Artikelnummer. 30356770
Change view
Klicka för att se tillgängliga alternativ
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Mængde
30356770 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30356770 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC984560500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemisk navn eller materiale N-Cyclohexyl-4-pentenamide
CAS 58839-90-0
Molekylær formel C11H19NO
Mængde 500 mg
Synonym N-Cyclohexyl-4-pentenamide (ACI)
InChI formel InChI=1S/C11H19NO/c1-2-3-9-11(13)12-10-7-5-4-6-8-10/h2,10H,1,3-9H2,(H,12,13)
SMIL C=CCCC(NC1CCCCC1)=O
IUPAC navn N-cyclohexylpent-4-enamide
Molekylvægt (g/mol) 181.27
Formel vægt 181.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.