missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexyl-4-fluorobenzamide, 97%, Thermo Scientific™

Catalog Number 15444088
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2342-50-9
C13H16FNO
221.28
MFCD00544070
STPFPZKUYANAOR-UHFFFAOYSA-N
benzamide, n-cyclohexyl-4-fluoro, n-cyclohexyl 4-fluorophenyl carboxamide
2305489
N-cyclohexyl-4-fluorobenzamide
FC1=CC=C(C=C1)C(=O)NC1CCCCC1
This item is not returnable. View return policy
2342-50-9
C13H16FNO
221.28
MFCD00544070
STPFPZKUYANAOR-UHFFFAOYSA-N
benzamide, n-cyclohexyl-4-fluoro, n-cyclohexyl 4-fluorophenyl carboxamide
2305489
N-cyclohexyl-4-fluorobenzamide
FC1=CC=C(C=C1)C(=O)NC1CCCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 221.27
Percent Purity 97%
Chemical Name or Material N-Cyclohexyl-4-fluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'