Learn More
Alfa Aesar™ N-Cyclohexyl-4-fluorobenzamide, 97%
Brand: Alfa Aesar™ H57640.03
Additional Details : CAS Number : 2342-50-9 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2342-50-9 | |
221.275 | |
STPFPZKUYANAOR-UHFFFAOYSA-N | |
2305489 | |
C1CCC(CC1)NC(=O)C2=CC=C(C=C2)F |
C13H16FNO | |
MFCD00544070 | |
benzamide, n-cyclohexyl-4-fluoro, n-cyclohexyl 4-fluorophenyl carboxamide | |
N-cyclohexyl-4-fluorobenzamide |
Specifications
N-Cyclohexyl-4-fluorobenzamide | |
2342-50-9 | |
MFCD00544070 | |
STPFPZKUYANAOR-UHFFFAOYSA-N | |
N-cyclohexyl-4-fluorobenzamide | |
2305489 | |
97% |
1g | |
C13H16FNO | |
benzamide, n-cyclohexyl-4-fluoro, n-cyclohexyl 4-fluorophenyl carboxamide | |
C1CCC(CC1)NC(=O)C2=CC=C(C=C2)F | |
221.275 | |
221.27 |