Få mere at vide
N-Cyanoacetylurethane, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Cyanoacetylurethane |
| CAS | 6629-04-5 |
| Molekylær formel | C6H8N2O3 |
| Mængde | 5 g |
| Synonym | N-(2-Cyanoacetyl)carbamic Acid Ethyl Ester, (Cyanoacetyl)carbamic Acid Ethyl Ester, Cyanoacetylurethan, Ethyl N-(cyanoacetyl)carbamate, Ethyl Cyanoacetylcarbamate, N-Carbethoxy-2-cyanoacetamide, N-Cyanoacetylurethane, NSC 59709 |
| InChI formel | InChI=1S/C6H8N2O3/c1-2-11-6(10)8-5(9)3-4-7/h2-3H2,1H3,(H,8,9,10) |
| SMIL | N#CCC(=O)NC(=O)OCC |
| IUPAC navn | ethyl N-(2-cyanoacetyl)carbamate |
| Molekylvægt (g/mol) | 156.14 |
| Formel vægt | 156.0535 |