Få mere at vide
N-Cyano-N'-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]-carbamimidic Acid Methyl Ester, TRC
Artikelnummer. 30363189
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363189 10 mg
30348890 50 mg
30356133 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30363189 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98806310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 138035-55-9
Molekylær formel C10 H15 N5 O S
Molekylvægt (g/mol) 253.32
Synonym Carbamimidic acid, N-cyano-N'-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]-, methyl ester (9CI), 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea, N-Cyano-N'-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]carbamimidic acid methyl ester, Cimetidine Hydrochloride Imp. B (EP)
IUPAC navn methyl N'-cyano-N-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]carbamimidate
SMIL COC(=NC#N)NCCSCc1nc[nH]c1C
Tekniske data
Kemisk navn eller materiale 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea
Mængde 10 mg
InChI formel InChI=1S/C10H15N5OS/c1-8-9(15-7-14-8)5-17-4-3-12-10(16-2)13-6-11/h7H,3-5H2,1-2H3,(H,12,13)(H,14,15)
Formel vægt 253.0997