Få mere at vide
N-Cyano-3-(cyanoamino)-N'-methyl-7,8-dithia-2,4,11-triazadodec-2-en-12-imidamide (Cimetidine Impurity), TRC
Artikelnummer. 30359934
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359934 250 mg
30352806 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359934 Leverandør Toronto Research Chemicals Leverandørnr. TRCC988275250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 74886-59-2
Molekylær formel C10 H18 N8 S2
Molekylvægt (g/mol) 314.43
Synonym 1,1'-(Disulphanediyldiethylene)bis(2-cyano-3-methylguanidine), Cimetidine Hydrochloride Imp. H (EP)
IUPAC navn 2-cyano-1-[2-[2-[(N'-cyano-N-methylcarbamimidoyl)amino]ethyldisulfanyl]ethyl]-3-methylguanidine
SMIL CNC(=NC#N)NCCSSCCNC(=NC#N)NC
Tekniske data
Kemisk navn eller materiale 1,1'-(Disulfanediyldiethylene)bis(2-cyano-3-methylguanidine)
Mængde 250 mg
InChI formel InChI=1S/C10H18N8S2/c1-13-9(17-7-11)15-3-5-19-20-6-4-16-10(14-2)18-8-12/h3-6H2,1-2H3,(H2,13,15,17)(H2,14,16,18)
Formel vægt 314.1096