Få mere at vide
N-Cinnamylglycine, TRC
Artikelnummer. 30363587
Skift visning
Mængde:
25 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363587 25 mg
30363531 10 g
30360270 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30363587 Leverandør Toronto Research Chemicals Leverandørnr. TRCC44212025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16534-24-0
Molekylær formel C11 H11 N O3
Molekylvægt (g/mol) 205.21
Synonym Glycine, N-(1-oxo-3-phenyl-2-propen-1-yl)-, Glycine, N-(1-oxo-3-phenyl-2-propenyl)- (9CI), Glycine, N-cinnamoyl- (6CI,8CI), N-(1-Oxo-3-phenyl-2-propen-1-yl)glycine, Cinnamoylglycine, N-Cinnamoylglycine, N-Cinnamylglycine, 2-Cinnamamidoacetic acid, 2-(3-Phenylacrylamido)acetic acid, 2-(3-Phenylprop-2-enamido)acetic acid, [(3-Phenylprop-2-enoyl)amino]acetic acid
IUPAC navn 2-(3-phenylprop-2-enoylamino)acetic acid
SMIL OC(=O)CNC(=O)C=Cc1ccccc1
Tekniske data
Kemisk navn eller materiale N-Cinnamylglycine
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C11H11NO3/c13-10(12-8-11(14)15)7-6-9-4-2-1-3-5-9/h1-7H,8H2,(H,12,13)(H,14,15)
Formel vægt 205.0739
Procent renhed >95%