Få mere at vide
N-Cbz-D-glutamic Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Cbz-D-glutamic Acid |
| CAS | 63648-73-7 |
| Molekylær formel | C11 H15 N O2 S |
| Mængde | 10 g |
| Synonym | N-[(Phenylmethoxy)carbonyl]-D-glutamic Acid, Benzyloxycarbonyl-D-glutamic Acid, N-(Benzyloxycarbonyl)-D-glutamic Acid, N-Carbobenzoxy-D-glutamic Acid, NSC 169149 |
| InChI formel | InChI=1S/C11H15NO2S/c1-7-3-4-9(5-8(7)2)15-6-10(12)11(13)14/h3-5,10H,6,12H2,1-2H3,(H,13,14)/t10-/m0/s1 |
| SMIL | Cc1ccc(SC[C@H](N)C(=O)O)cc1C |
| IUPAC navn | (2R)-2-amino-3-(3,4-dimethylphenyl)sulfanylpropanoic acid |
| Molekylvægt (g/mol) | 225.307 |
| Formel vægt | 225.082 |