Få mere at vide
N-(Carboxymethyl)-N-(phosphonomethyl)-glycine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(Phosphonomethyl)iminodiacetic acid monohydrate |
| CAS | 5994-61-6 |
| Molekylær formel | C5 H10 N O7 P . H2 O |
| Mængde | 1 g |
| Synonym | N-(Phosphonomethyl)iminodiacetic Acid, Acetic acid, [(phosphonomethyl)imino]di- (6CI,7CI,8CI), N-(Carboxymethyl)-N-(phosphonomethyl)glycine, Aminomethylphosphonic N,N-diacetic acid, MON 820, N-(Phosphonomethyl)iminodiacetic acid, Nitrilodiacetic acid methylphosphonic acid, Nitrilotris(methylene) triphonic acid, PMIDA, Phosphonomethyliminodiacetic acid |
| InChI formel | InChI=1S/C5H10NO7P/c7-4(8)1-6(2-5(9)10)3-14(11,12)13/h1-3H2,(H,7,8)(H,9,10)(H2,11,12,13) |
| SMIL | OC(=O)CN(CC(=O)O)CP(=O)(O)O |
| IUPAC navn | 2-[carboxymethyl(phosphonomethyl)amino]acetic acid |
| Molekylvægt (g/mol) | 245.12 |
| Formel vægt | 227.0195 |