Få mere at vide
N-(Carboxymethyl)-N,N-dimethyl-3-[[(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)sulfonyl]amino]-1-propanaminium, Inner Salt, TRC
Artikelnummer. 17929714
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17929714 25 mg
17947604 50 mg
17998614 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17929714 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17648025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34455-29-3
Molekylær formel C15 H19 F13 N2 O4 S
Molekylvægt (g/mol) 570.37
Synonym Capstone B, 1-Propanaminium, N-(carboxymethyl)-N,N-dimethyl-3-[[(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)sulfonyl]amino]-, hydroxide, inner salt, N-(Carboxymethyl)-N,N-dimethyl-3-[[(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)sulfonyl]amino]-1-propanaminium inner salt, N-(carboxymethyl)-N,N-dimethyl-3-((((perfluorohexyl)ethyl)sulfonyl)amino)-, hydroxide, inner salt, Capstone product B, 6:2 Fluorotelomer sulfonamide betaine, 6:2 FTAB
IUPAC navn 2-[dimethyl-[3-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctylsulfonylamino)propyl]azaniumyl]acetate
SMIL C[N+](C)(CCCNS(=O)(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)CC(=O)[O-]
Tekniske data
Kemisk navn eller materiale Capstone Product B
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C15H19F13N2O4S/c1-30(2,8-9(31)32)6-3-5-29-35(33,34)7-4-10(16,17)11(18,19)12(20,21)13(22,23)14(24,25)15(26,27)28/h29H,3-8H2,1-2H3
Formel vægt 570.0858
Procent renhed >95%