Få mere at vide
N-(Carboxymethyl)amphetamine Hydrochloride, TRC
Artikelnummer. 30367171
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367171 100 mg
30354055 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367171 Leverandør Toronto Research Chemicals Leverandørnr. TRCC181140100MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60342-93-0
Molekylær formel C11H15NO2•HCl
Molekylvægt (g/mol) 193.243646
Synonym N-(α-Methylphenethyl)glycine Hydrochloride
IUPAC navn 2-(1-phenylpropan-2-ylamino)acetic acid;hydrochloride
SMIL [H][C@]1(CN(C)CC2)N2C3=CC=CC=C3CC4=CC=CC=C14
Tekniske data
Kemisk navn eller materiale N-(Carboxymethyl)amphetamine Hydrochloride
Mængde 100 mg
InChI formel InChI=1S/C11H15NO2.ClH/c1-9(12-8-11(13)14)7-10-5-3-2-4-6-10;/h2-6,9,12H,7-8H2,1H3,(H,13,14);1H