Få mere at vide
N-Carbomethoxy-S-methylisothiourea, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Carbomethoxy-S-methylisothiourea |
| CAS | 39259-32-0 |
| Molekylær formel | C4H8N2O2S |
| Mængde | 25 mg |
| Synonym | Methyl N-(1-Amino-1-methylthiolmethylene)carbamate, N-Methoxycarbonyl-S-methylisothiourea, NSC 160239, [Imino(methylthio)methyl]-carbamic Acid Methyl Ester, S-Methyl-N-methoxycarbonylisothiourea |
| InChI formel | InChI=1S/C4H8N2O2S/c1-8-4(7)6-3(5)9-2/h1-2H3,(H2,5,6,7) |
| SMIL | COC(=O)NC(=N)SC |
| IUPAC navn | methyl N-(C-methylsulfanylcarbonimidoyl)carbamate |
| Molekylvægt (g/mol) | 148.18 |
| Formel vægt | 148.0306 |