missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzyloxy-D-proline, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 74.6°C to 74.3°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Specific Rotation Condition | + 40.20 (22.00°C c=2,alcohol) |
Specific Rotation | + 40.20 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'