Få mere at vide
N-Carbobenzyloxy-3-phenylthio-L-alanine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Carbobenzyloxy-3-phenylthio-L-alanine |
| CAS | 159453-24-4 |
| Molekylær formel | C17 H17 N O4 S |
| Mængde | 25 g |
| Synonym | L-Cysteine, S-phenyl-N-[(phenylmethoxy)carbonyl]-, S-Phenyl-N-[(phenylmethoxy)carbonyl]-L-cysteine, N-Benzyloxycarbonyl-S-phenyl-L-cysteine, N-Carbobenzoxy-S-phenyl-L-cysteine, N-Carbobenzoxy-S-phenylcysteine, N-CBZ-S-phenyl-L-cysteine, S-Phenyl-N-Cbz-L-cysteine, N-Cbz-Cys(Ph)-OH, (R)-2-[[(Benzyloxy)carbonyl]amino]-3-(phenylthio)propanoic acid, N-Carbobenzyloxy-3-phenylthio-L-alanine, N-[(Benzyloxy)carbonyl]-S-phenyl-L-cysteine, N-(Benzyloxycarbonyl)-S-phenyl-L-cysteine |
| InChI formel | InChI=1S/C17H17NO4S/c19-16(20)15(12-23-14-9-5-2-6-10-14)18-17(21)22-11-13-7-3-1-4-8-13/h1-10,15H,11-12H2,(H,18,21)(H,19,20)/t15-/m0/s1 |
| SMIL | OC(=O)[C@H](CSc1ccccc1)NC(=O)OCc2ccccc2 |
| IUPAC navn | (2R)-2-(phenylmethoxycarbonylamino)-3-phenylsulfanylpropanoic acid |
| Molekylvægt (g/mol) | 331.39 |
| Formel vægt | 331.0878 |