Få mere at vide
N-Carbobenzoxy-S-phenyl-L-cysteine Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Carbobenzoxy-S-phenyl-L-cysteine Methyl Ester |
| CAS | 153277-33-9 |
| Molekylær formel | C18H19NO4S |
| Mængde | 500 mg |
| Synonym | (2R)-2-[(Benzyloxycarbonyl)amino]-3-(phenylthio)propanoic Acid Methyl Ester, N-Benzyloxycarbonyl-S-phenyl-L-cysteine Methyl Ester, N-Carbobenzoxy-S-phenyl-L-cysteine Methyl Ester |
| InChI formel | InChI=1S/C18H19NO4S/c1-22-17(20)16(13-24-15-10-6-3-7-11-15)19-18(21)23-12-14-8-4-2-5-9-14/h2-11,16H,12-13H2,1H3,(H,19,21)/t16-/m0/s1 |
| SMIL | O=C(OC)[C@H](CSC1=CC=CC=C1)NC(OCC2=CC=CC=C2)=O |
| IUPAC navn | methyl (2R)-2-(phenylmethoxycarbonylamino)-3-phenylsulfanylpropanoate |
| Molekylvægt (g/mol) | 345.41 |
| Formel vægt | 345.1035 |